Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Carbamic acid, N-[[[(1,1-dimethylethoxy)carbonyl)amino]-1H-pyrazol-1-ylmethylene]-, 1,1-dimethylethyl ester | 152120-54-2 | 310.35 | 25 G
SDP

Catalog No. 5000449457
Encompass_Preferred

Carbamic acid, N-[[[(1,1-dimethylethoxy)carbonyl)amino]-1H-pyrazol-1-ylmethylene]-, 1,1-dimethylethyl ester, also referred to as N,N'-Di-Boc-1H-pyrazole-1-carboximidamide, is a chemical compound characterized by its molecular formula C14H22N4O4 and a molecular weight of 310.35. This product exhibits a high purity of 99.92% as determined by LCMS and typically appears as an off-white to light brown solid. It is recommended for research use only and has not been fully validated for medical applications.

  • Appearance: Off-white to light brown (solid)
  • Purity (LCMS): 99.92%
  • Water (KF): 0.02%
  • Recommended storage for powder: -20°C for 3 years; 4°C for 2 years
  • Recommended storage in solvent: -80°C for 6 months; -20°C for 1 month

Catalog No. 50-004-49457 Supplier Medchemexpress LLC Supplier No. 25GHYW002736
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Carbamic acid, N-[[[(1,1-dimethylethoxy)carbonyl)amino]-1H-pyrazol-1-ylmethylene]-, 1,1-dimethylethyl ester, also referred to as N,N'-Di-Boc-1H-pyrazole-1-carboximidamide, is a chemical compound characterized by its molecular formula C14H22N4O4 and a molecular weight of 310.35. This product exhibits a high purity of 99.92% as determined by LCMS and typically appears as an off-white to light brown solid. It is recommended for research use only and has not been fully validated for medical applications.

  • Appearance: Off-white to light brown (solid)
  • Purity (LCMS): 99.92%
  • Water (KF): 0.02%
  • Recommended storage for powder: -20°C for 3 years; 4°C for 2 years
  • Recommended storage in solvent: -80°C for 6 months; -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.