Learn More
Medchemexpress LLC Carbamic acid, N-[(1S)-1-[[(1-formyl-2-phenylethyl)amino]carbonyl]-2-methylpropyl]-, phenylmethyl | 88191-84-8 | 97.0% | 382.45 | 100 MG
SDP
Catalog No. 5000387614
Encompass_Preferred
MDL-28170 (Calpain Inhibitor III) is a potent, selective, and membrane-permeable cysteine protease inhibitor of calpain that rapidly penetrates the blood-brain barrier following systemic administration. It also blocks γ-secretase.
  • Potent, selective, and membrane-permeable cysteine protease inhibitor of calpain.
  • Rapidly penetrates the blood-brain barrier following systemic administration.
  • Blocks γ-secretase.
  • Significantly and time-dependently improves the recovery of synaptic responses in hippocampal slices following prolonged, moderate hypoxia without hypoxic depolarization (In Vitro).
  • Dose-dependently inhibits brain cysteine proteinase activity with an in vitro Ki of 0.01 μM.
Catalog No. 50-003-87614 Supplier Medchemexpress LLC Supplier No. HY18236100MG
MDL-28170 (Calpain Inhibitor III) is a potent, selective, and membrane-permeable cysteine protease inhibitor of calpain that rapidly penetrates the blood-brain barrier following systemic administration. It also blocks γ-secretase.
  • Potent, selective, and membrane-permeable cysteine protease inhibitor of calpain.
  • Rapidly penetrates the blood-brain barrier following systemic administration.
  • Blocks γ-secretase.
  • Significantly and time-dependently improves the recovery of synaptic responses in hippocampal slices following prolonged, moderate hypoxia without hypoxic depolarization (In Vitro).
  • Dose-dependently inhibits brain cysteine proteinase activity with an in vitro Ki of 0.01 μM.