Learn More
Medchemexpress LLC Carbamic acid, N-[(1S)-2-hydroxy-1-methylethyl]-, 1,1-dimethylethyl ester | 79069-13-9 | MFCD00043121 | 175.23 | 25 G
SDP
Catalog No. 5000448030
Encompass_Preferred
Boc-L-Alaninol is a chemical compound with the chemical name Carbamic acid, N-[(1S)-2-hydroxy-1-methylethyl]-, 1,1-dimethylethyl ester. It has a molecular formula of C8H17NO3 and a molecular weight of 175.23. This product appears as a white to off-white solid and boasts a purity of 99.8%.
  • White to off-white solid appearance
  • 1H NMR spectrum consistent with structure
  • Enantiomeric excess of 94.64%
  • Purity by GC: 99.8%
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
Catalog No. 50-004-48030 Supplier Medchemexpress LLC Supplier No. 25GHYW007729
Boc-L-Alaninol is a chemical compound with the chemical name Carbamic acid, N-[(1S)-2-hydroxy-1-methylethyl]-, 1,1-dimethylethyl ester. It has a molecular formula of C8H17NO3 and a molecular weight of 175.23. This product appears as a white to off-white solid and boasts a purity of 99.8%.
  • White to off-white solid appearance
  • 1H NMR spectrum consistent with structure
  • Enantiomeric excess of 94.64%
  • Purity by GC: 99.8%
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month