Learn More
Medchemexpress LLC Carbamic acid, N-[2-(2-aminoethoxy)ethyl]-, 1,1-dimethylethyl ester | 127828-22-2 | 204.27 | 25 G
SDP
Catalog No. 5000449915
Encompass_Preferred
Amino-PEG2-NH-Boc is a PEG-based PROTAC linker employed in the synthesis of Proteolysis-targeting Chimeras (PROTACs). PROTACs function by utilizing the intracellular ubiquitin-proteasome system to selectively degrade target proteins, featuring two ligands connected by a linker: one for an E3 ubiquitin ligase and the other for the target protein. This product is intended for research use only and has not been fully validated for medical applications.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Assists in the selective degradation of target proteins
  • Utilizes the intracellular ubiquitin-proteasome system
  • Purity of 98.0% (NMR)
  • Colorless to light yellow liquid appearance
Catalog No. 50-004-49915 Supplier Medchemexpress LLC Supplier No. 25GHYW008279
Amino-PEG2-NH-Boc is a PEG-based PROTAC linker employed in the synthesis of Proteolysis-targeting Chimeras (PROTACs). PROTACs function by utilizing the intracellular ubiquitin-proteasome system to selectively degrade target proteins, featuring two ligands connected by a linker: one for an E3 ubiquitin ligase and the other for the target protein. This product is intended for research use only and has not been fully validated for medical applications.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Assists in the selective degradation of target proteins
  • Utilizes the intracellular ubiquitin-proteasome system
  • Purity of 98.0% (NMR)
  • Colorless to light yellow liquid appearance