Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Carbamic acid, N-[2-[(2-aminoethyl)dithio]ethyl]-, 1,1-dimethylethyl ester | 485800-26-8 | 95.0% | 252.40 g/mol | C9H20N2O2S2 | 250 MG
SDP

Catalog No. 5000203863
Encompass_Preferred

Boc-Cystamine is a Boc-protected cystamine used as a cleavable disulfide-containing linker for the synthesis of antibody-drug conjugates (ADCs). It is intended for research use in linker synthesis and conjugation workflows, and releases payloads under reducing intracellular conditions.

  • Cleavable disulfide-containing linker for ADC synthesis.
  • Used in linker synthesis and conjugation workflows.
  • Purity 95.0%.
  • CAS number 485800-26-8.
  • Molecular weight 252.40 g/mol, formula C9H20N2O2S2.
  • Supplied as a solid; common packaging includes 250 mg.
  • Store protected from light and under inert atmosphere; in solvent: -80°C (6 months), -20°C (1 month).

Catalog No. 50-002-03863 Supplier Medchemexpress LLC Supplier No. HY140098250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boc-Cystamine is a Boc-protected cystamine used as a cleavable disulfide-containing linker for the synthesis of antibody-drug conjugates (ADCs). It is intended for research use in linker synthesis and conjugation workflows, and releases payloads under reducing intracellular conditions.

  • Cleavable disulfide-containing linker for ADC synthesis.
  • Used in linker synthesis and conjugation workflows.
  • Purity 95.0%.
  • CAS number 485800-26-8.
  • Molecular weight 252.40 g/mol, formula C9H20N2O2S2.
  • Supplied as a solid; common packaging includes 250 mg.
  • Store protected from light and under inert atmosphere; in solvent: -80°C (6 months), -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.