Learn More
Medchemexpress LLC Carbamic acid, N-[2-(2-propyn-1-yloxy)ethyl]-, 1,1-dimethylethyl ester. | 634926-63-9 | MFCD22418470 | 95.1% | 199.25 g/mol | C10H17NO3 | 5 G
SDP
Catalog No. 5000281792
Encompass_Preferred
O-Proparagyl-N-Boc-ethanolamine is a propargyl-functionalized, Boc-protected ethanolamine used as a crosslinker and synthetic building block. It provides an alkyne handle for copper-catalyzed azide-alkyne cycloaddition (click chemistry) and a t-Boc-protected amine that can be removed under mild acidic conditions to expose a free amine for further derivatization.
  • Alkyne handle for copper-catalyzed click chemistry enabling selective bioconjugation.
  • Boc-protected amine stable during handling and removable under mild acidic conditions.
  • Suitable as a crosslinker and synthetic intermediate in small-molecule and bioconjugation workflows.
  • Manufacturer COA reports GC purity 95.07% (rounded: 95.1%).
  • Recommended storage: pure form -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (up to 6 months).
Catalog No. 50-002-81792 Supplier Medchemexpress LLC Supplier No. HYW2595425G
O-Proparagyl-N-Boc-ethanolamine is a propargyl-functionalized, Boc-protected ethanolamine used as a crosslinker and synthetic building block. It provides an alkyne handle for copper-catalyzed azide-alkyne cycloaddition (click chemistry) and a t-Boc-protected amine that can be removed under mild acidic conditions to expose a free amine for further derivatization.
  • Alkyne handle for copper-catalyzed click chemistry enabling selective bioconjugation.
  • Boc-protected amine stable during handling and removable under mild acidic conditions.
  • Suitable as a crosslinker and synthetic intermediate in small-molecule and bioconjugation workflows.
  • Manufacturer COA reports GC purity 95.07% (rounded: 95.1%).
  • Recommended storage: pure form -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (up to 6 months).