Learn More
Medchemexpress LLC Carbamic acid, n-[2-[(r)-(3-chlorophenyl)...methyl ester, hydrochloride | 1264191-73-2 | MFCD19443251 | 98.2% | 561.54 | C26H42Cl2N4O5 | 10 MG
SDP
Catalog No. 5000298626
Encompass_Preferred
VTP-27999 Hydrochloride is the hydrochloride salt of an alkyl amine renin inhibitor supplied for preclinical research into hypertension and related end-organ diseases. Supplied as a purified solid with high aqueous solubility, it is intended for use in in vitro and in vivo studies.
  • High purity (≈98.2%) suitable for research applications.
  • Hydrochloride salt form for improved stability and handling.
  • Water solubility ≥ 100 mg/mL enabling convenient dosing and formulation.
  • Available in small package sizes for early-stage studies.
  • Store sealed and away from moisture to preserve stability.
Catalog No. 50-002-98626 Supplier Medchemexpress LLC Supplier No. HY7665210MG
VTP-27999 Hydrochloride is the hydrochloride salt of an alkyl amine renin inhibitor supplied for preclinical research into hypertension and related end-organ diseases. Supplied as a purified solid with high aqueous solubility, it is intended for use in in vitro and in vivo studies.
  • High purity (≈98.2%) suitable for research applications.
  • Hydrochloride salt form for improved stability and handling.
  • Water solubility ≥ 100 mg/mL enabling convenient dosing and formulation.
  • Available in small package sizes for early-stage studies.
  • Store sealed and away from moisture to preserve stability.