Learn More
Medchemexpress LLC Carbamic acid, N-[(2R,3S)-2-methyl-4-oxo-3-oxetanyl]-, 5-phenylpentyl ester | 1439366-88-7 | 99.7% | 1 ML
SDP
Catalog No. 5000231161
Encompass_Preferred
ARN 077 (enantiomer) is the less-active stereoisomer of ARN 077 (also reported as URB913 enantiomer). It is supplied for research use and reported to inhibit rat NAAA with an IC50 of 3.53 μM for the less-active enantiomer. The compound is offered as a 10 mM solution in DMSO (1 mL) and as solid quantities for laboratory use.
  • Supplied as 10 mM solution in DMSO, 1 mL, and as solids for flexibility in experiments.
  • High reported purity (≈99.7%), suitable for research applications.
  • Molecular formula C16H21NO4, molecular weight 291.34 g/mol.
  • Reported CAS number 1439366-88-7 for unambiguous identification.
  • Intended for research use only; not for human or diagnostic use.
Catalog No. 50-002-31161 Supplier Medchemexpress LLC Supplier No. HY120813A10MM 1ML
ARN 077 (enantiomer) is the less-active stereoisomer of ARN 077 (also reported as URB913 enantiomer). It is supplied for research use and reported to inhibit rat NAAA with an IC50 of 3.53 μM for the less-active enantiomer. The compound is offered as a 10 mM solution in DMSO (1 mL) and as solid quantities for laboratory use.
  • Supplied as 10 mM solution in DMSO, 1 mL, and as solids for flexibility in experiments.
  • High reported purity (≈99.7%), suitable for research applications.
  • Molecular formula C16H21NO4, molecular weight 291.34 g/mol.
  • Reported CAS number 1439366-88-7 for unambiguous identification.
  • Intended for research use only; not for human or diagnostic use.