Learn More
Medchemexpress LLC Carbamic acid, N-(3-aminobicyclo[1.1.1]pent-1-yl)-, 1,1-dimethylethyl ester | 1638767-25-5 | 100.0% | 198.27 g/mol | C10H18N2O2 | 25 G
SDP
Catalog No. 5000209312
Encompass_Preferred
tert-Butyl (3-aminobicyclo[1.1.1]pentan-1-yl)carbamate is a Boc-protected 3-aminobicyclo[1.1.1]pentane building block used as a drug intermediate in medicinal chemistry and synthetic applications.
  • Provides a tert-butyl carbamate (Boc) protected primary amine for selective deprotection and downstream functionalization.
  • High reported purity suitable for synthetic intermediates and reference compound use.
  • Solid, white to light yellow appearance for straightforward handling and storage.
  • Available in small to mid-scale quantities, including 25 g, to support research and development.
  • Stable under recommended storage: powder at -20°C for long-term storage or 4°C for shorter-term storage.
Catalog No. 50-002-09312 Supplier Medchemexpress LLC Supplier No. 25GHYW054424
tert-Butyl (3-aminobicyclo[1.1.1]pentan-1-yl)carbamate is a Boc-protected 3-aminobicyclo[1.1.1]pentane building block used as a drug intermediate in medicinal chemistry and synthetic applications.
  • Provides a tert-butyl carbamate (Boc) protected primary amine for selective deprotection and downstream functionalization.
  • High reported purity suitable for synthetic intermediates and reference compound use.
  • Solid, white to light yellow appearance for straightforward handling and storage.
  • Available in small to mid-scale quantities, including 25 g, to support research and development.
  • Stable under recommended storage: powder at -20°C for long-term storage or 4°C for shorter-term storage.