Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Carbamic acid, N-(3-aminopropyl)-, 1,1-dimethylethyl ester | 75178-96-0 | 99.8% | 174.24 | 50 G
SDP

Catalog No. 5000404751
Encompass_Preferred

tert-Butyl (3-aminopropyl)carbamate is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It appears as a colorless to off-white solid below 22°C and a liquid above 22°C. This product is for research use only and should be stored at 4°C, protected from light, and under nitrogen.

  • Biochemical reagent for life science research
  • Colorless to off-white appearance
  • Solid below 22°C, liquid above 22°C
  • Store at 4°C, protect from light, stored under nitrogen
  • Purity of 99.82%

Catalog No. 50-004-04751 Supplier Medchemexpress LLC Supplier No. HY4017250G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

tert-Butyl (3-aminopropyl)carbamate is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It appears as a colorless to off-white solid below 22°C and a liquid above 22°C. This product is for research use only and should be stored at 4°C, protected from light, and under nitrogen.

  • Biochemical reagent for life science research
  • Colorless to off-white appearance
  • Solid below 22°C, liquid above 22°C
  • Store at 4°C, protect from light, stored under nitrogen
  • Purity of 99.82%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.