Learn More
Medchemexpress LLC Carbamic acid, N-(4-bromobutyl)-, 1,1-dimethylethyl ester | 164365-88-2 | 252.15 | 10 G
SDP
Catalog No. 5000446455
Encompass_Preferred
Boc-NH-C4-Br is an alkyl chain-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker, one for an E3 ubiquitin ligase and the other for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Alkyl chain-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs utilize the ubiquitin-proteasome system
  • Target protein degradation
Catalog No. 50-004-46455 Supplier Medchemexpress LLC Supplier No. HYW00780310G
Boc-NH-C4-Br is an alkyl chain-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker, one for an E3 ubiquitin ligase and the other for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Alkyl chain-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs utilize the ubiquitin-proteasome system
  • Target protein degradation