Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Carbamic acid, N-(6-aminohexyl)-, 1,1-dimethylethyl ester | 51857-17-1 | 216.32 | 10 G
SDP

Catalog No. 5000449398
Encompass_Preferred

Carbamic acid, N-(6-aminohexyl)-, 1,1-dimethylethyl ester, also known as NH2-C6-NH-Boc, is a PROTAC linker with an alkyl/ether composition. It is utilized in the synthesis of Mcl-1 inhibitor based on PROTAC.

  • PROTAC linker with alkyl/ether composition
  • Utilized in the synthesis of Mcl-1 inhibitor based on PROTAC
  • Purity of 99.6%
  • Molecular weight of 216.32
  • Appearance is a colorless to light yellow solid-liquid mixture
  • Recommended storage at 4°C, protected from light
  • Solvent storage at -80°C for 6 months or -20°C for 1 month, protected from light
  • Soluble in ethanol (100 mg/mL with sonication)

Catalog No. 50-004-49398 Supplier Medchemexpress LLC Supplier No. HYW00829610G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Carbamic acid, N-(6-aminohexyl)-, 1,1-dimethylethyl ester, also known as NH2-C6-NH-Boc, is a PROTAC linker with an alkyl/ether composition. It is utilized in the synthesis of Mcl-1 inhibitor based on PROTAC.

  • PROTAC linker with alkyl/ether composition
  • Utilized in the synthesis of Mcl-1 inhibitor based on PROTAC
  • Purity of 99.6%
  • Molecular weight of 216.32
  • Appearance is a colorless to light yellow solid-liquid mixture
  • Recommended storage at 4°C, protected from light
  • Solvent storage at -80°C for 6 months or -20°C for 1 month, protected from light
  • Soluble in ethanol (100 mg/mL with sonication)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.