Learn More
Medchemexpress LLC Carbamic acid, N-(6-bromohexyl)-, 1,1-dimethylethyl ester | 142356-33-0 | MFCD06201019 | 98.0% | 280.20 g/mol | C11H22BrNO2 | 25 G
SDP
Catalog No. 5000202844
Encompass_Preferred
Boc-NH-C6-Br is a Boc-protected 6-bromohexyl linker used as a non-cleavable linker in antibody-drug conjugate (ADC) research. It features a Boc-protected primary amine for selective deprotection and a terminal bromide for nucleophilic substitution, enabling conjugation to payloads or scaffolds.
  • Provides a Boc-protected primary amine for controlled deprotection.
  • Contains a terminal bromide for alkylation or substitution reactions.
  • Functionally suited as a non-cleavable linker in ADC synthesis.
  • Supplied with high purity for research applications.
  • Available in multiple package sizes for scale flexibility.
  • Data sheet, COA, and SDS available for characterization and safety information.
Catalog No. 50-002-02844 Supplier Medchemexpress LLC Supplier No. HYW01156125G
Boc-NH-C6-Br is a Boc-protected 6-bromohexyl linker used as a non-cleavable linker in antibody-drug conjugate (ADC) research. It features a Boc-protected primary amine for selective deprotection and a terminal bromide for nucleophilic substitution, enabling conjugation to payloads or scaffolds.
  • Provides a Boc-protected primary amine for controlled deprotection.
  • Contains a terminal bromide for alkylation or substitution reactions.
  • Functionally suited as a non-cleavable linker in ADC synthesis.
  • Supplied with high purity for research applications.
  • Available in multiple package sizes for scale flexibility.
  • Data sheet, COA, and SDS available for characterization and safety information.