Learn More
Medchemexpress LLC Carbamic acid, N-methyl-N-[2-(methylamino)ethyl] ester, 2,2,2-trifluoroacetate | 1224601-17-5 | MFCD01938808 | 98.1% | 620.57 g/mol | C29H31F3N4O8 | 1 ML
SDP
Catalog No. 5000201969
Encompass_Preferred
SN-38-CO-DMEDA TFA is a research-grade carbamate intermediate (trifluoroacetate salt) used in the synthesis of antibody-drug conjugates such as LND1035. Supplied as a 10 mM solution in DMSO (1 mL), it has CAS number 1224601-17-5, chemical formula C29H31F3N4O8, molecular weight 620.57 g/mol, and reported purity 98.07%.
  • 10 mM solution in DMSO, 1 mL
  • Intended for use as a chemical synthesis intermediate
  • Reported purity 98.07%
  • CAS number 1224601-17-5
  • Available as solid quantities in multiple sizes
Catalog No. 50-002-01969 Supplier Medchemexpress LLC Supplier No. HY145767A10MM 1ML
SN-38-CO-DMEDA TFA is a research-grade carbamate intermediate (trifluoroacetate salt) used in the synthesis of antibody-drug conjugates such as LND1035. Supplied as a 10 mM solution in DMSO (1 mL), it has CAS number 1224601-17-5, chemical formula C29H31F3N4O8, molecular weight 620.57 g/mol, and reported purity 98.07%.
  • 10 mM solution in DMSO, 1 mL
  • Intended for use as a chemical synthesis intermediate
  • Reported purity 98.07%
  • CAS number 1224601-17-5
  • Available as solid quantities in multiple sizes