Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Carbamimidothioic acid, C,C'-(1,4-phenylenedi-2,1-ethanediyl) ester, hydrobromide (1:2) | 157254-60-9 | 98.0% | C12H20Br2N4S2 | 10MG
SDP

Catalog No. 5000260161
Encompass_Preferred

1,4-PBIT (dihydrobromide) is the dihydrobromide salt of the small-molecule 1,4-PBIT, supplied for research use. The compound is documented by manufacturer COA and SDS, and is provided as a white to off-white solid with molecular formula C12H20Br2N4S2, molecular weight 444.25, and reported purity 98.0%.

  • 98.0% purity as reported on the certificate of analysis
  • White to off-white solid form for solid-handling workflows
  • Molecular formula C12H20Br2N4S2 and molecular weight 444.25
  • Recommended storage: in solvent -80°C (6 months) or -20°C (1 month), sealed
  • Used as a nitric oxide synthase inhibitor in research applications

Catalog No. 50-002-60161 Supplier Medchemexpress LLC Supplier No. HY13522410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,4-PBIT (dihydrobromide) is the dihydrobromide salt of the small-molecule 1,4-PBIT, supplied for research use. The compound is documented by manufacturer COA and SDS, and is provided as a white to off-white solid with molecular formula C12H20Br2N4S2, molecular weight 444.25, and reported purity 98.0%.

  • 98.0% purity as reported on the certificate of analysis
  • White to off-white solid form for solid-handling workflows
  • Molecular formula C12H20Br2N4S2 and molecular weight 444.25
  • Recommended storage: in solvent -80°C (6 months) or -20°C (1 month), sealed
  • Used as a nitric oxide synthase inhibitor in research applications

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.