Learn More
Medchemexpress LLC Carbamodithioic acid, N,N-diethyl-, sodium salt (1:1) | 148-18-5 | 99.2% | 50 G
SDP
Supplier:  Medchemexpress LLC HYB1637A50G
Encompass_Preferred
Ditiocarb sodium is an orally active copper reagent that demonstrates various activities including antioxidation, chelation, anti-tumor effects, immunomodulation, and anti-HIV properties. It is suitable for use in research related to tumors, inflammatory, and immune-related diseases.
  • Exhibits activities such as antioxidation, chelation, anti-tumor effects, immunomodulation, and anti-HIV properties.
  • Can be used in the research of tumors, inflammatory, and immune-related diseases.
  • Acts as an accelerator in the cementation of copper from copper sulfate solution on zinc.
  • Increases the rate of copper cementation by reacting with Cu2+ solution to form a copper diethyldithiocarbamate complex.
  • Can inhibit cisplatin-induced nephrotoxicity and gastrointestinal toxicity in rats.
  • Exhibits antitumor activity in a mouse skin tumor model.
Catalog No. 50-004-12722