Learn More
AdipoGen Carbamoylcholine chloride (1 g)
SDP
Supplier:  AdipoGen AGCR13649G001
Encompass_Preferred
Chemical. CAS: 51-83-2. Formula: C6H15N2O2 . Cl. MW: 182.7. Acetylcholine analog that activates acetylcholine receptors (AChR). Non-selective agonist of nicotinic (nAChR) and muscarinic (mAChR) receptors that is resistant to the action of cholinesterases. Used to study responses mediated by nAChR and mAChR, including smooth muscle contraction, gut motility and neuronal signaling. Shown to inhibit apoptotic death of cultured neurons.
Catalog No. 50-205-4023