Learn More
Medchemexpress LLC Carbamoyl phosphate dilithium salt | 1866-68-8 | 152.89 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY160610A100MG
Encompass_Preferred
Carbamoyl phosphate dilithium salt is an antisickling agent that plays a significant role in the urea cycle. It functions by carbamylating the amino-terminal valine residues of hemoglobin S in erythrocytes, which helps interfere with the sickling phenomenon. This product exhibits toxicity due to the presence of the lithium cation and is intended for research use only.
  • Functions as an antisickling agent.
  • Plays a role in the urea cycle.
  • Carbamylates amino-terminal valine residues of hemoglobin S.
  • Helps interfere with the sickling phenomenon in erythrocytes.
  • Intended for research use only.
Catalog No. 50-003-94566