Learn More
Medchemexpress LLC Carbamoyl phosphate (dilithium salt) | 1866-68-8 | 152.89 | 25 MG
SDP
Catalog No. 5000394432
Encompass_Preferred
Carbamoyl phosphate dilithium salt is an antisickling agent that plays a significant role in the urea cycle. It carbamylates the amino-terminal valine residues of hemoglobin S in erythrocytes, which interferes with the sickling phenomenon. However, it exhibits toxicity due to the lithium cation.
  • Antisickling agent
  • Plays a significant role in the urea cycle
  • Carbamylates the amino-terminal valine residues of hemoglobin S in erythrocytes
  • Interferes with the sickling phenomenon
  • Exhibits toxicity due to the lithium cation
Catalog No. 50-003-94432 Supplier Medchemexpress LLC Supplier No. 25MGHY160610A
Carbamoyl phosphate dilithium salt is an antisickling agent that plays a significant role in the urea cycle. It carbamylates the amino-terminal valine residues of hemoglobin S in erythrocytes, which interferes with the sickling phenomenon. However, it exhibits toxicity due to the lithium cation.
  • Antisickling agent
  • Plays a significant role in the urea cycle
  • Carbamylates the amino-terminal valine residues of hemoglobin S in erythrocytes
  • Interferes with the sickling phenomenon
  • Exhibits toxicity due to the lithium cation