Learn More
Medchemexpress LLC Carbamoyl phosphate (dilithium salt) | 1866-68-8 | 152.89 | 50 MG
SDP
Catalog No. 5000394567
Encompass_Preferred
Carbamoyl phosphate dilithium salt is an antisickling agent that plays a significant role in the urea cycle. It carbamylates the amino-terminal valine residues of hemoglobin S in erythrocytes, which interferes with the sickling phenomenon. This compound is for research use only and not sold to patients.
  • Acts as an antisickling agent
  • Plays a significant role in the urea cycle
  • Carbamylates amino-terminal valine residues of hemoglobin S in erythrocytes
  • Interferes with the sickling phenomenon
  • For research use only
Catalog No. 50-003-94567 Supplier Medchemexpress LLC Supplier No. HY160610A50MG
Carbamoyl phosphate dilithium salt is an antisickling agent that plays a significant role in the urea cycle. It carbamylates the amino-terminal valine residues of hemoglobin S in erythrocytes, which interferes with the sickling phenomenon. This compound is for research use only and not sold to patients.
  • Acts as an antisickling agent
  • Plays a significant role in the urea cycle
  • Carbamylates amino-terminal valine residues of hemoglobin S in erythrocytes
  • Interferes with the sickling phenomenon
  • For research use only