Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Carbamoyl phosphate dilithium salt | 1866-68-8 | MFCD00038144 | 152.89 g/mol | CH2Li2NO5P | 10 MG
SDP

Catalog No. 5000285464
Encompass_Preferred

Carbamoyl phosphate dilithium salt is an inorganic biochemical reagent used in research applications, including enzymatic assays and studies of the urea cycle. It is supplied in milligram-scale quantities for laboratory use and is intended for research use only (not for human or clinical use).

  • used as a reagent in urea cycle and enzymatic assays
  • commonly supplied in milligram quantities for bench experiments
  • CAS number 1866-68-8 and molecular weight 152.89 g/mol
  • chemical formula CH2Li2NO5P
  • provided with supporting documentation such as SDS and COA (refer to supplier documentation for handling and purity details)

Catalog No. 50-002-85464 Supplier Medchemexpress LLC Supplier No. HY160610A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Carbamoyl phosphate dilithium salt is an inorganic biochemical reagent used in research applications, including enzymatic assays and studies of the urea cycle. It is supplied in milligram-scale quantities for laboratory use and is intended for research use only (not for human or clinical use).

  • used as a reagent in urea cycle and enzymatic assays
  • commonly supplied in milligram quantities for bench experiments
  • CAS number 1866-68-8 and molecular weight 152.89 g/mol
  • chemical formula CH2Li2NO5P
  • provided with supporting documentation such as SDS and COA (refer to supplier documentation for handling and purity details)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.