Learn More
Cayman Chemical Carbamoylcholine (chloride), Purity: ≥98%, CAS: 51-83-2, 5 g
SDP
Catalog No. 502973340
Encompass_Preferred
An agonist of both nAChR and mAChR receptors, with reported Ki values ranging from 10 to 10,000 nM for different receptors and different preparations; used to study responses mediated by nAChR and mAChR, including smooth muscle contraction, gut motility, and neuronal signaling
Catalog No. 50-297-3340 Supplier Cayman Chemical Supplier No. 144865 G
An agonist of both nAChR and mAChR receptors, with reported Ki values ranging from 10 to 10,000 nM for different receptors and different preparations; used to study responses mediated by nAChR and mAChR, including smooth muscle contraction, gut motility, and neuronal signaling