Learn More
AdipoGen Carbamoylcholine chloride
SDP
Supplier:  AdipoGen CDXC0306G001
Encompass_Preferred
Chemical. CAS 51-83-2. Formula C6H15N2O2 . Cl. MW 182.65. Non-selective cholinergic agonist that is resistant to the action of cholinesterases inhibits apoptotic death of cultured neurons, and may induce apoptosis in thymocytes. Carbamoylcholine, also known as carbachol, is an analog of acetylcholine that activates acetylcholine receptors AChR. Carbamoylcholine is an agonist of both nicotinic nAChR and muscarinic mAChR receptors.
Catalog No. 50-256-1394