Learn More
Medchemexpress LLC Carbamyl-PAF (C-PAF) | 91575-58-5 | MFCD00065891 | 99.0% | 538.70 | C26H55N2O7P | 5 MG
SDP
Catalog No. 5000222802
Encompass_Preferred
Carbamyl-PAF (C-PAF) is a platelet-activating factor analogue used as a research reagent to probe PAF receptor signaling and related cellular responses. It is provided as a high-purity ethanol solution for biochemical and cell-based assays and requires low-temperature storage.
  • High purity: 99.0%.
  • Supplied as solution in ethanol for convenient dilution.
  • Stable when stored at -20°C for up to 2 years.
  • Suitable for receptor pharmacology and cellular assays.
  • Molecular weight 538.70; formula C26H55N2O7P.
Catalog No. 50-002-22802 Supplier Medchemexpress LLC Supplier No. HY1372575MG
Carbamyl-PAF (C-PAF) is a platelet-activating factor analogue used as a research reagent to probe PAF receptor signaling and related cellular responses. It is provided as a high-purity ethanol solution for biochemical and cell-based assays and requires low-temperature storage.
  • High purity: 99.0%.
  • Supplied as solution in ethanol for convenient dilution.
  • Stable when stored at -20°C for up to 2 years.
  • Suitable for receptor pharmacology and cellular assays.
  • Molecular weight 538.70; formula C26H55N2O7P.