Learn More
Cayman Chemical Carbamylated Bovine Serum Albumin, Purity: ≥85% estimated by SDS-PAGE, 1 mg
SDP
Catalog No. 502983890
Encompass_Preferred
Source: Albumin isolated from bovine plasma and modified with potassium cyanate • MW: 69.3 kDa
Catalog No. 50-298-3890 Supplier Cayman Chemical Supplier No. 210751 MG
Source: Albumin isolated from bovine plasma and modified with potassium cyanate • MW: 69.3 kDa