Learn More
Cayman Chemical Carbamylated Core HIstonsb
SDP
Catalog No. 502984193
Encompass_Preferred
Source: Histones from calf thymus modified with potassium cyanate
Catalog No. 50-298-4193 Supplier Cayman Chemical Supplier No. 21076100 UG
Source: Histones from calf thymus modified with potassium cyanate