Learn More
Cayman Chemical CarbamylatIon IP KIt 1ea
SDP
Catalog No. 502896252
Encompass_Preferred
A cognate dye ligand for the Pepper RNA aptamer Upon binding to the Pepper RNA aptamer the Pepper-HBC 530 complex displays excitation/emission maxima of 485/530 nm respectively has been used in the live cell imaging of RNA
Catalog No. 50-289-6252 Supplier Cayman Chemical Supplier No. 6019301 EA
A cognate dye ligand for the Pepper RNA aptamer Upon binding to the Pepper RNA aptamer the Pepper-HBC 530 complex displays excitation/emission maxima of 485/530 nm respectively has been used in the live cell imaging of RNA