Learn More
Cayman Chemical CarbaprostacyclIn-bIotIn 100ug
SDP
Catalog No. 502968136
Encompass_Preferred
An affinity probe which allows carbaprostacyclin to be detected in complexes with transmembrane receptors and/or nucleic or protein binding partners
Catalog No. 50-296-8136 Supplier Cayman Chemical Supplier No. 18213100 UG
An affinity probe which allows carbaprostacyclin to be detected in complexes with transmembrane receptors and/or nucleic or protein binding partners