Learn More
Medchemexpress LLC Carbaryl-d7 | 362049-56-7 | 99.0% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB1315S11MG
Encompass_Preferred
Carbaryl-d7 is a deuterium-labeled form of Carbaryl, an acetylcholinesterase inhibitor that prevents the breakdown of acetylcholine in the synaptic cleft, leading to its accumulation and causing neurotoxicity. It can also function as an insecticide.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-13585