Learn More
Cayman Chemical Carbazochrome, Purity: ≥98%, CAS: 69-81-8, 50 g
SDP
Catalog No. 502949908
Encompass_Preferred
A hemostatic agent; reduces blood loss by 45% in a mouse model of traumatic hemorrhage at 0.1 mg/kg; topical administration induces blood coagulation at back wound sites in mice
Catalog No. 50-294-9908 Supplier Cayman Chemical Supplier No. 3295150 G
A hemostatic agent; reduces blood loss by 45% in a mouse model of traumatic hemorrhage at 0.1 mg/kg; topical administration induces blood coagulation at back wound sites in mice