Learn More
Medchemexpress LLC Carbazochrome | 69-81-8 | 99.9% | 250 MG
SDP
Catalog No. 5000413953
Encompass_Preferred
Carbazochrome is a capillary stabiliser and antihemorrhagic agent used for the research of haemorrhage.
  • Inhibits Bradykinin-induced and thrombin-induced formation of [3H]IP3 in a concentration-dependent manner.
  • Suppresses enhancement of permeability induced by vasoactive substances.
  • Inhibits severe pulmonary dysfunction induced by the intravenous injection of radiographic contrast media.
  • Attenuates pulmonary dysfunction induced by a radiographic contrast medium in rats.
Catalog No. 50-004-13953 Supplier Medchemexpress LLC Supplier No. 250MGHYB1587
Carbazochrome is a capillary stabiliser and antihemorrhagic agent used for the research of haemorrhage.
  • Inhibits Bradykinin-induced and thrombin-induced formation of [3H]IP3 in a concentration-dependent manner.
  • Suppresses enhancement of permeability induced by vasoactive substances.
  • Inhibits severe pulmonary dysfunction induced by the intravenous injection of radiographic contrast media.
  • Attenuates pulmonary dysfunction induced by a radiographic contrast medium in rats.