Learn More
Medchemexpress LLC Carbazochrome (sodium sulfonate) | 51460-26-5 | 98.0% | 100 MG
SDP
Catalog No. 5000409479
Encompass_Preferred
Carbazochrome sodium sulfonate (AC-17) is a capillary stabilizer and an antihemorrhagic agent for hemorrhage research.
  • Inhibits Bradykinin-induced and thrombin-induced formation of [3H]IP3
  • Suppresses permeability enhancement induced by vasoactive substances
  • Attenuates pulmonary dysfunction induced by radiographic contrast media
  • For research use only
Catalog No. 50-004-09479 Supplier Medchemexpress LLC Supplier No. HYB0491A100MG
Carbazochrome sodium sulfonate (AC-17) is a capillary stabilizer and an antihemorrhagic agent for hemorrhage research.
  • Inhibits Bradykinin-induced and thrombin-induced formation of [3H]IP3
  • Suppresses permeability enhancement induced by vasoactive substances
  • Attenuates pulmonary dysfunction induced by radiographic contrast media
  • For research use only