Learn More
Medchemexpress LLC Carbazochrome sodium sulfonate | 51460-26-5 | MFCD00867493 | 98.0% | 1 ML
SDP
Catalog No. 5000406416
Encompass_Preferred
Carbazochrome sodium sulfonate (AC-17) is a capillary stabiliser and an antihemorrhagic agent used for the research of haemorrhage.
  • Used for research of haemorrhage
  • Acts as a capillary stabiliser
  • Functions as an antihemorrhagic agent
  • Inhibits Bradykinin-induced and thrombin-induced formation of [3H]IP3
  • Suppresses enhancement of permeability induced by vasoactive substances
  • Attenuates pulmonary dysfunction induced by radiographic contrast media
Catalog No. 50-004-06416 Supplier Medchemexpress LLC Supplier No. HYB0491A10MM/1ML
Carbazochrome sodium sulfonate (AC-17) is a capillary stabiliser and an antihemorrhagic agent used for the research of haemorrhage.
  • Used for research of haemorrhage
  • Acts as a capillary stabiliser
  • Functions as an antihemorrhagic agent
  • Inhibits Bradykinin-induced and thrombin-induced formation of [3H]IP3
  • Suppresses enhancement of permeability induced by vasoactive substances
  • Attenuates pulmonary dysfunction induced by radiographic contrast media