Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Carbazochrome (sodium sulfonate) | 51460-26-5 | MFCD00867493 | 98.0% | 25 MG
SDP

Catalog No. 5000450639
Encompass_Preferred

Carbazochrome sodium sulfonate (AC-17) is a capillary stabilizer and antihemorrhagic agent used in research on hemorrhage. It inhibits Bradykinin-induced and thrombin-induced [3H]IP3 formation, and also suppresses the enhancement of permeability induced by vasoactive substances.

  • Capillary stabilizer
  • Antihemorrhagic agent
  • Inhibits Bradykinin-induced and thrombin-induced [3H]IP3 formation
  • Suppresses enhanced permeability induced by vasoactive substances
  • Inhibits pulmonary dysfunction caused by radiographic contrast media
  • Attenuates pulmonary dysfunction in rats
  • Recommended storage at 4°C, sealed and away from moisture
  • Soluble in DMSO (≥ 49 mg/mL) and water (6.67 mg/mL)
  • Appearance: Yellow to orange solid

Catalog No. 50-004-50639 Supplier Medchemexpress LLC Supplier No. 25MGHYB0491A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Carbazochrome sodium sulfonate (AC-17) is a capillary stabilizer and antihemorrhagic agent used in research on hemorrhage. It inhibits Bradykinin-induced and thrombin-induced [3H]IP3 formation, and also suppresses the enhancement of permeability induced by vasoactive substances.

  • Capillary stabilizer
  • Antihemorrhagic agent
  • Inhibits Bradykinin-induced and thrombin-induced [3H]IP3 formation
  • Suppresses enhanced permeability induced by vasoactive substances
  • Inhibits pulmonary dysfunction caused by radiographic contrast media
  • Attenuates pulmonary dysfunction in rats
  • Recommended storage at 4°C, sealed and away from moisture
  • Soluble in DMSO (≥ 49 mg/mL) and water (6.67 mg/mL)
  • Appearance: Yellow to orange solid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.