Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbazochrome 99.0+%, TCI America™
SDP

Catalog No. A017625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A017625G 25 g
A01765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A017625G Supplier TCI America Supplier No. A017625G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 69-81-8
Molecular Formula C10H12N4O3
Molecular Weight (g/mol) 236.231
MDL Number MFCD00059705
InChI Key XSXCZNVKFKNLPR-UHFFFAOYSA-N
Synonym Adrenochrome Semicarbazone
PubChem CID 5478929
IUPAC Name [(3-hydroxy-1-methyl-6-oxo-2,3-dihydroindol-5-ylidene)amino]urea
SMILES CN1CC(C2=CC(=NNC(=O)N)C(=O)C=C21)O

Specifications

Melting Point 203°C
Quantity 25 g
Formula Weight 236.23
Percent Purity ≥99.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Carbazochrome
RTECSNumber NM1925500
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.