Learn More
Medchemexpress LLC Carbazole-9-ethanol | 1484-14-6 | MFCD00518704 | 99.9% | 211.26 | C14H13NO | 5 G
SDP
Catalog No. 5000304081
Encompass_Preferred
Carbazole-9-ethanol is an organic research reagent (C14H13NO, MW 211.26, CAS 1484-14-6) used as a building block in synthetic chemistry and life-science research. It appears as a white to off-white solid and is supplied in small laboratory pack sizes with high reported purity, making it suitable for method development and compound synthesis applications.
  • High reported purity for reliable synthetic and analytical results.
  • Useful building block for the synthesis of carbazole derivatives.
  • Suitable for medicinal chemistry and life-science research workflows.
  • Available in multiple small pack sizes for laboratory-scale work.
Catalog No. 50-003-04081 Supplier Medchemexpress LLC Supplier No. HYW0143345G
Carbazole-9-ethanol is an organic research reagent (C14H13NO, MW 211.26, CAS 1484-14-6) used as a building block in synthetic chemistry and life-science research. It appears as a white to off-white solid and is supplied in small laboratory pack sizes with high reported purity, making it suitable for method development and compound synthesis applications.
  • High reported purity for reliable synthetic and analytical results.
  • Useful building block for the synthesis of carbazole derivatives.
  • Suitable for medicinal chemistry and life-science research workflows.
  • Available in multiple small pack sizes for laboratory-scale work.