Learn More
Medchemexpress LLC Carbazole-9-ethanol | 1484-14-6 | MFCD00518704 | 99.9% | 211.26 g/mol | C14H13NO | 10 G

Supplier: Medchemexpress LLC HYW01433410G
Carbazole-9-ethanol is an organic carbazole derivative used as a research reagent in chemical and life-science laboratories. It serves as a building block or intermediate in organic synthesis and biochemical studies, and is supplied with documentation to support laboratory use.
- High purity (99.93%).
- Molecular formula C14H13NO.
- Molecular weight 211.26 g/mol.
- Available as a 10 g supply for bench-scale work.
- Provided with datasheet, certificate of analysis, and safety data sheet.
- For research use only; not intended for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.