Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbazole, 96%, Thermo Scientific™

Catalog No. AC108265000
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC108265000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC108265000 Supplier Thermo Scientific Chemicals Supplier No. 108265000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86-74-8
Molecular Formula C12H9N
Molecular Weight (g/mol) 167.21
MDL Number MFCD00004960
InChI Key UJOBWOGCFQCDNV-UHFFFAOYSA-N
Synonym carbazole, dibenzopyrrole, diphenylenimine, 9-azafluorene, diphenylenimide, diphenyleneimine, dibenzo b,d pyrrole, usaf ek-600, unii-0p2197hhhn
PubChem CID 6854
ChEBI CHEBI:27543
IUPAC Name 9H-carbazole
SMILES N1C2=C(C=CC=C2)C2=C1C=CC=C2

Specifications

Melting Point 240.0°C to 246.0°C
Boiling Point 355.0°C
Flash Point 220°C
Quantity 500 g
Solubility Information Solubility in water: insoluble. Other solubilities: slightly soluble in acetic acid - soluble in h2so4, 1g/3mL quinoline - 1g/6mL pyridine - 1g/35mL ether, 1g/9mL acetone - 1g/120mL benzene, 1g/135mL abs. alcohol - slightly soluble in, petroleum ether and
Formula Weight 167.21
Chemical Name or Material Carbazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.