Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbazole Potassium Salt 90.0+%, TCI America™
SDP

Catalog No. C071425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C071425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C071425G Supplier TCI America Supplier No. C071425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6033-87-0
Molecular Formula C12H8KN
Molecular Weight (g/mol) 205.301
MDL Number MFCD00070509
InChI Key WTSKDGZCVMJGDJ-UHFFFAOYSA-N
Synonym Potassium Carbazole
PubChem CID 12261362
IUPAC Name potassium;carbazol-9-ide
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3[N-]2.[K+]

Specifications

Quantity 25 g
Formula Weight 205.30
Percent Purity ≥90.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Carbazole Potassium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.