Learn More
Cayman Chemical Carbazomycin B, Purity: ≥95%, CAS: 75139-38-7, 5 mg
SDP
Catalog No. 502946863
Encompass_Preferred
A bacterial metabolite with diverse biological activities; active against a panel of seven fungi (MICs = 3.2-200 µg/ml) and a panel of seven bacteria (MICs = 25-50 µg/ml), as well as P. falciparum and C. albicans (IC50s = 2.37 and 19.6 µg/ml, respectively); active against a panel of five plant-pathogenic fungi (MICs = 12.5-200 µg/ml); cytotoxic to MCF-7, KB, NCI H187, and Vero cells (IC50s = 8.4, 8.6, 4.2, and 48.9 µM, respectively); inhibits 5-LO activity in RBL-1 cell extracts (IC50 = 1.5 µM)
Catalog No. 50-294-6863 Supplier Cayman Chemical Supplier No. 330895 MG
A bacterial metabolite with diverse biological activities; active against a panel of seven fungi (MICs = 3.2-200 µg/ml) and a panel of seven bacteria (MICs = 25-50 µg/ml), as well as P. falciparum and C. albicans (IC50s = 2.37 and 19.6 µg/ml, respectively); active against a panel of five plant-pathogenic fungi (MICs = 12.5-200 µg/ml); cytotoxic to MCF-7, KB, NCI H187, and Vero cells (IC50s = 8.4, 8.6, 4.2, and 48.9 µM, respectively); inhibits 5-LO activity in RBL-1 cell extracts (IC50 = 1.5 µM)