Learn More
Cayman Chemical Carbendazim, Purity: ≥98%, CAS: 10605-21-7, 100 mg
SDP
Catalog No. 502993689
Encompass_Preferred
A carbamate fungicide and an active metabolite of benomyl and thiophanate-methyl; a degradation product of benomyl; active against N. parvum, B. dothidea, D. seriata, and L. theobromae in vitro (EC50s = 0.01-0.07 mg/L) and protects grapevines from pruning wounds caused by D. seriata or D. mutila in vivo; inhibits the growth of, and microtubule polymerization in, S. cerevisiae at 100 µM; impairs meiosis and steroidogenesis in rat seminiferous tubules ex vivo and increases prostate weight in rats at 100 mg/kg; has been found as a contaminant in various jellies and jams and in wastewater influent and effluent
Catalog No. 50-299-3689 Supplier Cayman Chemical Supplier No. 23852100 MG
A carbamate fungicide and an active metabolite of benomyl and thiophanate-methyl; a degradation product of benomyl; active against N. parvum, B. dothidea, D. seriata, and L. theobromae in vitro (EC50s = 0.01-0.07 mg/L) and protects grapevines from pruning wounds caused by D. seriata or D. mutila in vivo; inhibits the growth of, and microtubule polymerization in, S. cerevisiae at 100 µM; impairs meiosis and steroidogenesis in rat seminiferous tubules ex vivo and increases prostate weight in rats at 100 mg/kg; has been found as a contaminant in various jellies and jams and in wastewater influent and effluent