Learn More
Medchemexpress LLC HY-13582 100mg Medchemexpress, Carbendazim CAS:10605-21-7 Purity:>98%
SDP
Catalog No. 502081405
Encompass_Preferred
Medchemexpress, HY-13582 100mg Carbendazim CAS:10605-21-7 Carbendazim is a potent and orally active broad-spectrum benzimidazole fungicide and can be acts as a pesticide for fungal diseases research, such as Seproria, Fusarium and Sclerotina. Carbendazim is a benzimidazole (HY-Y1825) derivative with antitumor activity and used for cancer research, especially advanced solid tumors and lymphoma. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1405 Supplier Medchemexpress LLC Supplier No. HY13582100MG
Medchemexpress, HY-13582 100mg Carbendazim CAS:10605-21-7 Carbendazim is a potent and orally active broad-spectrum benzimidazole fungicide and can be acts as a pesticide for fungal diseases research, such as Seproria, Fusarium and Sclerotina. Carbendazim is a benzimidazole (HY-Y1825) derivative with antitumor activity and used for cancer research, especially advanced solid tumors and lymphoma. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.