Learn More
Medchemexpress LLC Carbendazim-d4 | 291765-95-2 | 99.6% | C9H5D4N3O2 | 10MG
SDP
Catalog No. 5000261347
Encompass_Preferred
Carbendazim-d4 is the deuterium-labeled analogue of the fungicide carbendazim, supplied as a 10 mg reference standard for analytical and research use. It is intended for use as an isotopically labeled internal standard or tracer in quantitative mass spectrometry and related analytical workflows, offering consistent performance and storage stability under recommended conditions.
  • High chemical and isotopic purity.
  • Suitable as an internal standard for mass spectrometry.
  • Provided as a stable, dry powder for long-term storage.
  • Compatible with common organic solvents for sample preparation.
  • Characterized molecular weight and formula for analytical identification.
Catalog No. 50-002-61347 Supplier Medchemexpress LLC Supplier No. HY13582S10MG
Carbendazim-d4 is the deuterium-labeled analogue of the fungicide carbendazim, supplied as a 10 mg reference standard for analytical and research use. It is intended for use as an isotopically labeled internal standard or tracer in quantitative mass spectrometry and related analytical workflows, offering consistent performance and storage stability under recommended conditions.
  • High chemical and isotopic purity.
  • Suitable as an internal standard for mass spectrometry.
  • Provided as a stable, dry powder for long-term storage.
  • Compatible with common organic solvents for sample preparation.
  • Characterized molecular weight and formula for analytical identification.