Learn More
Medchemexpress LLC Carbendazim-d4 (deuterium-labeled carbendazim) | 291765-95-2 | MFCD04112602 | 99.6% | 195.21 | C9H5D4N3O2 | 5 MG
SDP
Catalog No. 5000223082
Encompass_Preferred
Carbendazim-d4 is a deuterium-labeled stable-isotope reference standard used for analytical and mass spectrometry applications, providing a reliable internal standard for quantitative analysis, method validation, and metabolite identification.
  • Isotopically labeled (d4) internal standard for mass spectrometry.
  • High chemical purity and isotopic enrichment for accurate quantitation.
  • Available in small milligram quantities ideal for reference standards.
  • Stable powder form with recommended cold storage for long-term retention of quality.
  • Suitable for method development, validation, and metabolite studies.
Catalog No. 50-002-23082 Supplier Medchemexpress LLC Supplier No. HY13582S5MG
Carbendazim-d4 is a deuterium-labeled stable-isotope reference standard used for analytical and mass spectrometry applications, providing a reliable internal standard for quantitative analysis, method validation, and metabolite identification.
  • Isotopically labeled (d4) internal standard for mass spectrometry.
  • High chemical purity and isotopic enrichment for accurate quantitation.
  • Available in small milligram quantities ideal for reference standards.
  • Stable powder form with recommended cold storage for long-term retention of quality.
  • Suitable for method development, validation, and metabolite studies.