Learn More
Teknova 100mg/mL Carbenicillin. 50mL, Sterile.
SDP
Catalog No. NC9619768
Encompass
Premade antibiotic solutions are ready-to-use as a supplement for your tissue culture, bacteria media or other specific needs. Antibiotics are often used in cell culture to prevent contamination, maintain aseptic conditions, or select for cells containing genetic modifications.
This item is not returnable. View return policy
Catalog No. NC9619768 Supplier Teknova Supplier No. C2199
Premade antibiotic solutions are ready-to-use as a supplement for your tissue culture, bacteria media or other specific needs. Antibiotics are often used in cell culture to prevent contamination, maintain aseptic conditions, or select for cells containing genetic modifications.