Learn More
IBI SCIENTIFIC 3P Carbenicillin Disodium Salt | 4800-94-6 | MFCD00077683 | 25 g
Catalog No. NC0753434
Encompass_Preferred
Carbenicillin - 25gm Carbenicillin is an antibiotic which inhibits bacterial cell wall synthesis, activity against gram-positive and gram-negative bacteria, and for cell culture media applications. Recommended use is 100mg/ml.
Catalog No. NC0753434 Supplier IBI SCIENTIFIC 3P Supplier No. IB02025
Carbenicillin - 25gm Carbenicillin is an antibiotic which inhibits bacterial cell wall synthesis, activity against gram-positive and gram-negative bacteria, and for cell culture media applications. Recommended use is 100mg/ml.