Learn More
Medchemexpress LLC HY-B0525A 1g , Carbenicillin (disodium) CAS:4800-94-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0525A1G
Encompass_Preferred
HY-B0525A 1g Carbenicillin (disodium) CAS: 4800-94-6 Carbenicillin disodium is a beta-lactam penicillin derivative that interference with final stage of bacterial cell wall synthesis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0295