Learn More
Gold Biotechnology Inc Carbenicillin (Disodium), USP Grade 250 g
SDP
Catalog No. NC1655000
Encompass
Carbenicillin is a member of the penicillin β-lactam antibiotics. However, unlike most β-lactams, carbenicillin disodium is limited to primarily gram-negative bacteria including Pseudomonas aeruginosa and common enteric species. It can be used in selection protocols and is often preferred over ampicillin due to its increased stability. This allows for less contamination of unwanted bacterial colonies. In addition, carbenicillin has been used to regulate bacterial growth in plants, yielding low regeneration frequencies. It can be used in genetic transformation applications to select for AmpR transformed cells.
Catalog No. NC1655000 Supplier Gold Biotechnology Inc Supplier No. C103250
Carbenicillin is a member of the penicillin β-lactam antibiotics. However, unlike most β-lactams, carbenicillin disodium is limited to primarily gram-negative bacteria including Pseudomonas aeruginosa and common enteric species. It can be used in selection protocols and is often preferred over ampicillin due to its increased stability. This allows for less contamination of unwanted bacterial colonies. In addition, carbenicillin has been used to regulate bacterial growth in plants, yielding low regeneration frequencies. It can be used in genetic transformation applications to select for AmpR transformed cells.