Learn More
Ubpbio Carbenicillin disodium salt, CAS# 4800-94-6, > 95% purity
Catalog No. 502339320
Encompass_Preferred
Carbenicillin is a semi-synthetic bactericidal antibiotic in the penicillin family. It primarily acts on Gram-negative bacteria. Carbenicillin is more stable than ampicillin and results in fewer satellite colonies on selection plates.
Catalog No. 50-233-9320 Supplier Ubpbio Supplier No. P802025
Carbenicillin is a semi-synthetic bactericidal antibiotic in the penicillin family. It primarily acts on Gram-negative bacteria. Carbenicillin is more stable than ampicillin and results in fewer satellite colonies on selection plates.