Learn More
Indofine Chemical Carbenicillin Disodium, 5 Gm
SDP
Catalog No. 50908430
Encompass_Preferred
Molecular Formula: C17H16N2O6SNa2, Purity 92%, Molecular Weight: 422.38, Appearance: White powder, Storage conditions: Store at 0-8C
Catalog No. 50-908-430 Supplier Indofine Chemical Supplier No. C0109 5 GM
Molecular Formula: C17H16N2O6SNa2, Purity 92%, Molecular Weight: 422.38, Appearance: White powder, Storage conditions: Store at 0-8C